Please login to your online account to display your discounted pricing

Mouse anti-CD45RO, Clone: SPM125, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD45RO
Clone SPM125
Concentration 0.2mg/mL
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23293901

 
novus biologicals canada
NBP23293901ML
0.1mL Each for $139.00

NBP23293910

 
novus biologicals canada
NBP23293910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD45RO
Clone SPM125
Concentration 0.2mg/mL
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting