Please login to your online account to display your discounted pricing

Mouse anti-CD45RO, Clone: SPM125, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293910

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Primate
Antigen CD45RO
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P08575
Host Species Mouse
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
Isotype IgG2a, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Clone SPM125
Concentration 0.2mg/mL
Conjugate Unlabeled
Reconstitution Protein A purified