Please login to your online account to display your discounted pricing

Mouse anti-CD45RO, Clone: SPM125, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293901

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Primate
Antigen CD45RO
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P08575
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Isotype IgG2a, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone SPM125
Concentration 0.2mg/mL
Conjugate Unlabeled