Please login to your online account to display your discounted pricing

Mouse anti-CD45RO, Clone: SPM125, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473501

  • / Each
 

Description & Specifications

Specifications

Antigen CD45RO
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.
Clone SPM125
Conjugate Unlabeled
Cross Reactivity Chimpanzee
Cross Reactivity Human
Gene Accession No. P08575
Host Species Mouse
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Concentration 1mg/mL