Please login to your online account to display your discounted pricing

Mouse anti-CD45RO, Clone: SPM125, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473502

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Chimpanzee
Cross Reactivity Human
Antigen CD45RO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.
Reconstitution Protein A purified
Clone SPM125
Concentration 1mg/mL
Conjugate Unlabeled
Gene Accession No. P08575
Immunogen Cultured human T-cells from an IL-2-dependent T-cell line (CA1)