Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: SPM569, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD45RB
Clone SPM569
Concentration 0.2mg/mL
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23293801

 
novus biologicals canada
NBP23293801ML
0.1mL Each for $139.00

NBP23293810

 
novus biologicals canada
NBP23293810ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD45RB
Clone SPM569
Concentration 0.2mg/mL
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting