Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: SPM569, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA.
Gene Accession No. P08575
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Format Purified
Conjugate Unlabeled
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL
Host Species Mouse
Antigen CD45RB
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Clone SPM569
Concentration 0.2mg/mL
Purification Method Purified
Reconstitution Protein A purified
Primary or Secondary Primary