Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: SPM569, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480302

  • / Each
 

Description & Specifications

Specifications

Reconstitution Protein A purified
Antigen CD45RB
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08575
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM569
Conjugate Unlabeled
Cross Reactivity Human
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Isotype IgG1, κ
Gene ID (Entrez) 5788
Concentration 1mg/mL