missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: PD7/26, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD45RB
Clone PD7/26
Concentration 1mg/mL
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Gene Accession No. P08575
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23470801 Novus Biologicals Canada
NBP23470801MG
0.1mg Each for $384.00
NBP23470802 Novus Biologicals Canada
NBP23470802MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD45RB
1mg/mL
P08575
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
PD7/26
Human peripheral blood lymphocytes maintained in T cell growth factor
5788
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot