Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: PD7/26, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470801

  • / Each
 

Description & Specifications

Specifications

Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Cross Reactivity Human
Antigen CD45RB
Format Purified
Gene Accession No. P08575
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone PD7/26
Concentration 1mg/mL
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Primary or Secondary Primary
Gene ID (Entrez) 5788