Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: BRA-11/BRA-11G, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD45RB
Clone BRA-11/BRA-11G
Concentration 1mg/mL
Immunogen Non-T
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23456401

 
novus biologicals canada
NBP23456401MG
0.1mg Each for $384.00

NBP23456402

 
novus biologicals canada
NBP23456402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD45RB
Clone BRA-11/BRA-11G
Concentration 1mg/mL
Immunogen Non-T
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot