Please login to your online account to display your discounted pricing

Mouse anti-CD45RB, Clone: BRA-11/BRA-11G, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456401

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Immunogen Non-T
Host Species Mouse
Conjugate Unlabeled
Format Purified
Clone BRA-11/BRA-11G
Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 5788
Gene Accession No. P08575
Concentration 1mg/mL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Rat (Negative)
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Antigen CD45RB
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Reconstitution Protein A purified
Regulatory Status RUO