Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: SPM568, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293610

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 5788
Antigen CD45RA
Format Purified
Gene Accession No. P08575
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Regulatory Status RUO
Clone SPM568
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Purification Method Purified
Quantity 1.0mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified