Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: SPM568, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P08575
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Purified
Regulatory Status RUO
Cross Reactivity Human
Antigen CD45RA
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Clone SPM568
Concentration 0.2mg/mL