Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: SPM568, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Antigen CD45RA
Clone SPM568
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.1mg
Gene ID (Entrez) 5788
Host Species Mouse
Primary or Secondary Primary
Regulatory Status RUO
Reconstitution Protein A purified
Gene Accession No. P08575
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL