Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: JS-83, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD45RA
Clone JS-83
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143701

 
novus biologicals canada
NBP143701100TESTS
100 Tests Each for $296.00

NBP14370125

 
novus biologicals canada
NBP14370125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD45RA
Clone JS-83
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry