Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: JS-83, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14311301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Immunogen The immunogen for this antibody was CD45RA.
Clone JS-83
Format Affinity Purified
Antigen CD45RA
Concentration 0.2mg/mL
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 5788
Purification Method Affinity Purified
Formulation Phosphate buffer pH 7.2
Isotype IgG1, κ
Applications Flow Cytometry