Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: HI100, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD45RA
Clone HI100
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14341001

 
novus biologicals canada
NBP14341001MG
0.1mg Each for $285.00

NBP14341003

 
novus biologicals canada
NBP1434100025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD45RA
Clone HI100
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)