Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: HI100, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14341001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14341001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation Phosphate buffer pH 7.2
Conjugate Unlabeled
Format Affinity Purified
Antigen CD45RA
Regulatory Status RUO
Immunogen The immunogen for this antibody was CD45RA.
Clone HI100
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Concentration 0.5mg/mL
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG2b, κ
Gene ID (Entrez) 5788
Purification Method Affinity Purified