Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: HI100, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434100025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14341003

  • / Each
 

Description & Specifications

Specifications

Antigen CD45RA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone HI100
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Murine
Immunogen The immunogen for this antibody was CD45RA.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Target Species Human