Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: HI100, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $428.00

Specifications

Antigen CD45RA
Clone HI100
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142850

 
novus biologicals canada
NBP142850100TESTS
100 Tests Each for $428.00

NBP14285025

 
novus biologicals canada
NBP14285025TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD45RA
Clone HI100
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD45RA.
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry