Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: 158-4D3, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen CD45RA
Clone 158-4D3
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes were used as immunogen to generate the CD45RA antibody.
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21519301

 
novus biologicals canada
NBP21519301MG
0.1mg Each for $329.00

NBP21519302

 
novus biologicals canada
NBP21519302MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen CD45RA
Clone 158-4D3
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes were used as immunogen to generate the CD45RA antibody.
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot