Please login to your online account to display your discounted pricing

Mouse anti-CD45RA, Clone: 158-4D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519302

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD45RA
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Quantity 0.2mg
Conjugate Unlabeled
Gene Accession No. P08575
Immunogen Stimulated human leukocytes were used as immunogen to generate the CD45RA antibody.
Isotype IgG2a, κ
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO) (4th Leucocyte Typing Workshop: Code No. N31). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas. Reacts with Chimpanzee, Common Marmoset.
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone 158-4D3
Concentration 0.2mg/mL