Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM570, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293510

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Quantity 1.0mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P08575
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM570
Concentration 0.2mg/mL
Primary or Secondary Primary