Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM570, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Antigen CD45
Formulation PBS with 0.05% BSA.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P08575
Reconstitution Protein A purified
Regulatory Status RUO
Host Species Mouse
Quantity 0.1mL
Format Purified
Isotype IgG1, κ
Primary or Secondary Primary
Clone SPM570
Concentration 0.2mg/mL
Purification Method Purified
Gene ID (Entrez) 5788
Immunogen Isolated neoplastic cells from T cell lymphoma