Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM570, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480401

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Cross Reactivity Human
Antigen CD45
Format Purified
Gene Accession No. P08575
Immunogen Isolated neoplastic cells from T cell lymphoma
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled
Purification Method Purified
Regulatory Status RUO
Clone SPM570