Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM570, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23480402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23480402

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Regulatory Status RUO
Host Species Mouse
Antigen CD45
Gene ID (Entrez) 5788
Primary or Secondary Primary
Reconstitution Protein A purified
Gene Accession No. P08575
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Isolated neoplastic cells from T cell lymphoma
Conjugate Unlabeled
Format Purified
Isotype IgG1, κ
Clone SPM570
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.