Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM569 + SPM570, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23438305ML

 View all options for this product


For Research Use Only

Catalog No. NBP23438305


Description & Specifications


Antigen CD45
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM569 + SPM570
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P08575
Host Species Mouse
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity Recognizes the CD45 leukocyte common antigen (LCA) family which is comprised of at least four isoforms of membrane glycoproteins (220, 205, 190, 180kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues.
Reconstitution Protein A purified