Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM569 + SPM570, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443002

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08575
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 5788
Test Specificity Recognizes the CD45 leukocyte common antigen (LCA) family which is comprised of at least four isoforms of membrane glycoproteins (220, 205, 190, 180kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues.
Clone SPM569 + SPM570
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human