Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: SPM569 + SPM570, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443001

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Cross Reactivity Human
Gene ID (Entrez) 5788
Isotype IgG1, κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Format Purified
Concentration 1mg/mL
Primary or Secondary Primary
Gene Accession No. P08575
Reconstitution Protein A purified
Host Species Mouse
Quantity 0.1mg
Antigen CD45
Conjugate Unlabeled
Clone SPM569 + SPM570
Test Specificity Recognizes the CD45 leukocyte common antigen (LCA) family which is comprised of at least four isoforms of membrane glycoproteins (220, 205, 190, 180kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues.
Regulatory Status RUO