Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: HI30, PE-Cy7, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen CD45
Clone HI30
Concentration 0.005mg/mL
Immunogen This antibody was made against an amino acid sequence that is conserved among all CD45 isoforms.
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142956

 
novus biologicals canada
NBP142956100TESTS
100 Tests Each for $329.00

NBP14295625

 
novus biologicals canada
NBP14295625TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen CD45
Clone HI30
Concentration 0.005mg/mL
Immunogen This antibody was made against an amino acid sequence that is conserved among all CD45 isoforms.
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Murine
Isotype IgG1
Conjugate PE-Cy7
Format Purified
Purification Method Purified
Applications Flow Cytometry