Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: HI30, Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14297925TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14297925

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Applications Flow Cytometry
Clone HI30
Concentration 0.2mg/mL
Conjugate Cy5
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Mouse
Immunogen This antibody was made against an amino acid sequence that is conserved among all CD45 isoforms.
Isotype IgG1
Purification Method Purified
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788