Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: HI30, Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142979100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142979

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Applications Flow Cytometry
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Host Species Mouse
Isotype IgG1
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Clone HI30
Conjugate Cy5
Formulation Phosphate buffer pH 7.2. IgG purified.
Immunogen This antibody was made against an amino acid sequence that is conserved among all CD45 isoforms.
Quantity 100 Tests
Concentration 0.2mg/mL
Purification Method Purified