Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: 2B11 + PD7/26, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD45
Clone 2B11 + PD7/26
Concentration 0.2mg/mL
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23428701

 
novus biologicals canada
NBP23428701ML
0.1mL Each for $139.00

NBP23428705

 
novus biologicals canada
NBP23428705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD45
Clone 2B11 + PD7/26
Concentration 0.2mg/mL
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting