Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: 2B11 + PD7/26, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428701

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P08575
Regulatory Status RUO
Antigen CD45
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone 2B11 + PD7/26
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Isotype IgG1, κ
Gene ID (Entrez) 5788