Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: 2B11 + PD7/26, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428705

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Format Purified
Gene Accession No. P08575
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Isotype IgG1, κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Clone 2B11 + PD7/26
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Host Species Mouse
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Cross Reactivity Human