Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: 2B11 + PD7/26, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452801

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
Conjugate Unlabeled
Antigen CD45
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08575
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Clone 2B11 + PD7/26
Concentration 1mg/mL
Cross Reactivity Human