Please login to your online account to display your discounted pricing

Mouse anti-CD45, Clone: 2B11, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452701

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Clone 2B11
Antigen CD45
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Isolated neoplastic cells from T cell lymphoma
Regulatory Status RUO
Concentration 1mg/mL
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08575
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Reconstitution Protein A purified
Conjugate Unlabeled