Please login to your online account to display your discounted pricing

Mouse anti-CD45.2, Clone: 104, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen CD45.2
Clone 104
Concentration 0.5mg/mL
Immunogen B10.S mouse thymocytes and splenocytes
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364101

 
novus biologicals canada
NBP14364101MG
0.1mg Each for $197.00

NBP14364105

 
novus biologicals canada
NBP143641005MG
0.05mg Each for $131.00

NBP1436415

 
novus biologicals canada
NBP14364105MG
0.5mg Each for $395.00
Description & Specifications

Specifications

Antigen CD45.2
Clone 104
Concentration 0.5mg/mL
Immunogen B10.S mouse thymocytes and splenocytes
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)