Please login to your online account to display your discounted pricing

Mouse anti-CD45.1, Clone: A20, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen CD45.1
Clone A20
Concentration 0.5mg/mL
Immunogen Mouse CD 45.1 (Leukocyte Common Antigen)
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364201

 
novus biologicals canada
NBP14364201MG
0.1mg Each for $197.00

NBP14364205

 
novus biologicals canada
NBP143642005MG
0.05mg Each for $131.00

NBP1436425

 
novus biologicals canada
NBP14364205MG
0.5mg Each for $395.00
Description & Specifications

Specifications

Antigen CD45.1
Clone A20
Concentration 0.5mg/mL
Immunogen Mouse CD 45.1 (Leukocyte Common Antigen)
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry