Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM544, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437301

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Antigen CD44
Isotype IgG2a, κ
Gene Accession No. P16070
Test Specificity Recognizes a cell surface glycoprotein of 80-95kDa (CD44) on lymphocytes, monocytes, and granulocytes. Its epitope is resistant to digestion by trypsin and chymotrypsin. Reacts with Baboon, Green Monkey.
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Host Species Mouse
Immunogen Stimulated human leukocytes
Reconstitution Protein A purified
Gene ID (Entrez) 960
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM544
Purification Method Purified
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Format Purified
Conjugate Unlabeled