Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM544, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437305

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.5mL
Regulatory Status RUO
Immunogen Stimulated human leukocytes
Concentration 0.2mg/mL
Gene Accession No. P16070
Primary or Secondary Primary
Antigen CD44
Host Species Mouse
Conjugate Unlabeled
Format Purified
Gene ID (Entrez) 960
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG2a, κ
Clone SPM544
Test Specificity Recognizes a cell surface glycoprotein of 80-95kDa (CD44) on lymphocytes, monocytes, and granulocytes. Its epitope is resistant to digestion by trypsin and chymotrypsin. Reacts with Baboon, Green Monkey.
Reconstitution Protein A purified
Formulation PBS with 0.05% BSA.