missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM544, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD44
Clone SPM544
Concentration 1mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P16070
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23442001 Novus Biologicals Canada
NBP23442001MG
0.1mg Each for $384.00
NBP23442002 Novus Biologicals Canada
NBP23442002MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD44
1mg/mL
P16070
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM544
Stimulated human leukocytes
960
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot