Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM544, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442002

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Host Species Mouse
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Clone SPM544
Cross Reactivity Human
Cross Reactivity Primate
Antigen CD44
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P16070
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a cell surface glycoprotein of 80-95kDa (CD44) on lymphocytes, monocytes, and granulocytes. Its epitope is resistant to digestion by trypsin and chymotrypsin. Reacts with Baboon, Green Monkey.
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled