Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM544, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Primate
Antigen CD44
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P16070
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Test Specificity Recognizes a cell surface glycoprotein of 80-95kDa (CD44) on lymphocytes, monocytes, and granulocytes. Its epitope is resistant to digestion by trypsin and chymotrypsin. Reacts with Baboon, Green Monkey.
Reconstitution Protein A purified
Clone SPM544
Concentration 1mg/mL
Conjugate Unlabeled