Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM521, Novus Biologicals™

$268.00 - $518.93

Specifications

Antigen CD44
Clone SPM521
Immunogen Synthetic peptide corresponding to human CD44.
Gene ID (Entrez) 960
Formulation 10mM PBS/1% BSA buffer pH 7.4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13554901

 
novus biologicals canada
NBP13554901ML
0.1mL Each for $268.00

NBP13554910

 
novus biologicals canada
NBP13554910ML
1mL Each for $518.93

NBP13554970

 
novus biologicals canada
NBP13554970ML
7mL Each for $397.00
Description & Specifications

Specifications

Antigen CD44
Clone SPM521
Immunogen Synthetic peptide corresponding to human CD44.
Gene ID (Entrez) 960
Formulation 10mM PBS/1% BSA buffer pH 7.4
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting