Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM521, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13554970ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13554970

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Quantity 7mL
Isotype IgG2a
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Formulation 10mM PBS/1% BSA buffer pH 7.4
Cross Reactivity Human
Gene ID (Entrez) 960
Antigen CD44
Purification Method Purified
Immunogen Synthetic peptide corresponding to human CD44.
Host Species Mouse
Format Purified
Test Specificity This product is specific for CD44
Clone SPM521
Reconstitution Protein A purified
Conjugate Unlabeled