Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM521, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13554901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13554901

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen CD44
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG2a
Clone SPM521
Format Purified
Monoclonal or Polyclonal Monoclonal
Immunogen Synthetic peptide corresponding to human CD44.
Gene ID (Entrez) 960
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity This product is specific for CD44
Storage Requirements Store at 4°C. Do not freeze.
Formulation 10mM PBS/1% BSA buffer pH 7.4
Quantity 0.1mL
Purification Method Purified
Reconstitution Protein A purified
Regulatory Status RUO