Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: SPM521, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13554910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13554910

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Human
Antigen CD44
Format Purified
Formulation 10mM PBS/1% BSA buffer pH 7.4
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide corresponding to human CD44.
Isotype IgG2a
Purification Method Purified
Quantity 1mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Test Specificity This product is specific for CD44
Reconstitution Protein A purified
Conjugate Unlabeled
Clone SPM521