Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: 156-3C11, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD44
Clone 156-3C11
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P16070
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23432701

 
novus biologicals canada
NBP23432701ML
0.1mL Each for $139.00

NBP23432705

 
novus biologicals canada
NBP23432705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD44
Clone 156-3C11
Concentration 0.2mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P16070
Gene ID (Entrez) 960
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Baboon
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting